CELLvo™ human Mesenchymal Stem Cells- Wharton's Jelly (hMSC-WJ), StemBioSys, Inc. (SBS)
WJ-500-XF-T75 WJ-500-XF-T150 WJ-500-XF-6WP WJ-500-000
76200-216EA 655.64 USD
76200-216 76200-332 76200-334 76200-330
CELLvo™ human Mesenchymal Stem Cells- Wharton's Jelly (hMSC-WJ), StemBioSys, Inc. (SBS)
CELLvo™ human Mesenchymal Stem Cells-Wharton's Jelly are adult multipotent stem cells cryopreserved at passage 2.
  • Highly proliferative
  • Smaller cell size
  • xeno-free
  • Higher percentage of SSEA4+ cells
Wharton's Jelly is a rich source of highly proliferative, multipotent, mesenchymal stem cells. These cells are known to be highly immunomodulatory and be capable of trilineage differentiation.
CELLvo™ human Mesenchymal Stem Cells- Wharton's Jelly have been isolated and cultured under xeno free conditions on CELLvo™ XF-Matrix. CELLvo™ MSC-WJ proliferate more quickly, have a smaller cell size and show a higher percentage of SSEA4 (+) cells as compared to Wharton's Jellly MSCs isolated and cultured on other substrates.
Información para pedidos: CELLvo™ human Mesenchymal Stem Cell - Wharton's Jelly (hMSC-WJ) are derived from a single donor and cryopreserved at P2. CELLvo™ hMSC-WJ can be ordered as a single vial of cells or as a kit to include CELLvo™ XF Matrix.
Envase: CELLvo™ hMSC-WJ arrive on dry ice and may be purchased to include a sleeve of CELLvo™ XF Matrix. hMSC-WJ should be stored in liquid nitrogen vapor upon arrival. CELLvo™ XF Matrix should be stored at 2-8° upon arrival. Avoid extended exposure to light.
Precaución: CELLvo™ products are not for human use. These products should not be used for diagnostic or therapeutic purposes. Handle in accordance with established bio-safety practices.