Metro® Stationary Space Saver Worktables, Stainless Steel
Supplier: Metro International
WTS2436FS WTS2436US WTS2460US WTS2448FS WTS2448US WTS2460FS
76181-320EA 2243.61 USD
76181-320 76181-326 76181-284 76181-322 76181-328 76181-324
Metro® Stationary Space Saver Worktables, Stainless Steel
Furniture Tables
Maximize every inch with Metro® Stationary Space Saver Worktables.
  • Durable stainless steel components
  • Available with a solid bottom shelf or 3-sided frame
  • Easy assembly
Made from durable stainless steel components to stand up to the harshest applications and environments, sturdy construction provides a robust 800 lbs. load rating.
The 1" diameter posts and patented Super Erecta Shelf construction assure a rigid, sway-free table. Heavy-duty 14-gauge stainless steel virtually eliminates any "drumming."
Easily assembles in minutes without the use of special tools.