Venturi V25 LX Series General Purpose Bench Fume Hoods with 28" Vertical Rising Sash, Type 316L Stainless Steel Liner, Kewaunee Scientific
Supplier: Kewaunee
V25F283696LS V25F282472LM V25F283648LS V25F282496LN V25F283648LM V25F283648LN V25F283048LS V25F283072LN V25F283096LN V25F282448LN V25F282496LM V25F282460LM V25F283096LM V25F283048LM V25F283660LM V25F282460LS V25F283060LM V25F283072LM V25F283672LS V25F283096LS V25F283060LN V25F282496LS V25F282472LN V25F283660LS V25F283072LS V25F283048LN V25F282472LS V25F282448LM V25F283672LN V25F283672LM V25F282448LS V25F283696LM V25F283060LS V25F283660LN V25F283696LN V25F282460LN
KSV25F283060LNEA 23530.6 USD
KSV25F283060LN KSV25F282472LM KSV25F283648LS KSV25F282496LM KSV25F282472LN KSV25F282496LN KSV25F283060LM KSV25F282472LS KSV25F283048LM KSV25F283048LN KSV25F283648LM KSV25F283648LN KSV25F283060LS KSV25F283672LM KSV25F283672LN KSV25F282448LN KSV25F282448LM KSV25F283696LM KSV25F283696LN KSV25F282448LS KSV25F283072LN KSV25F282460LM KSV25F283096LM KSV25F283096LN KSV25F283072LM KSV25F282460LN KSV25F282460LS KSV25F283696LS KSV25F283672LS KSV25F283096LS KSV25F283072LS KSV25F283660LM KSV25F283660LN KSV25F283660LS KSV25F283048LS KSV25F282496LS
Venturi V25 LX Series General Purpose Bench Fume Hoods with 28" Vertical Rising Sash, Type 316L Stainless Steel Liner, Kewaunee Scientific
Fume Hoods and Enclosures Chemical Fume Hoods
The Supreme Air Venturi provides the safest user-friendly fume hood manufactured today.
  • Un paralleled Venturi enhanced containment technology
  • High performance/energy efficient
  • Unique ultra-smooth sash system
  • Exceptional viewing height
  • Flush airfoil
  • LED lights with color and intensity adjustability
  • Electromechanical sash stops
The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features, including: adjustable LED lights, electromechanical sash stops, ultra-smooth sash system, and push button sash management.
Certificaciones: UL and cUL listed. Tested to ASHRAE 110, NIH03-112C, ASTM D552, and NFPA 45.
Información para pedidos: Work top and base cabinets not included with fume hood.