Venturi V25 LX Series General Purpose Bench Fume Hoods with 28" Vertical Rising Sash, Kemglass Liner, Kewaunee Scientific
Supplier: Kewaunee
V25F283660GS V25F282448GM V25F283060GS V25F282460GM V25F283048GN V25F282496GM V25F283096GS V25F283696GN V25F283696GS V25F283696GM V25F283072GM V25F283648GM V25F282472GS V25F283096GN V25F282496GN V25F282448GN V25F282448GS V25F282460GS V25F283648GN V25F283672GS V25F283096GM V25F283048GM V25F283072GN V25F283060GM V25F282472GN V25F283648GS V25F282496GS V25F282472GM V25F283060GN V25F283660GM V25F283672GM V25F282460GN V25F283660GN V25F283672GN V25F283072GS V25F283048GS
KSV25F282460GMEA 18996.05 USD
KSV25F282460GM KSV25F282460GN KSV25F283696GN KSV25F283096GN KSV25F283672GN KSV25F283072GS KSV25F283696GM KSV25F283072GM KSV25F283696GS KSV25F283096GM KSV25F283672GS KSV25F283072GN KSV25F283048GM KSV25F283660GN KSV25F283048GN KSV25F283660GM KSV25F282496GN KSV25F282496GM KSV25F282472GS KSV25F283048GS KSV25F282496GS KSV25F283648GM KSV25F283648GN KSV25F283648GS KSV25F282472GM KSV25F282472GN KSV25F283060GS KSV25F283060GM KSV25F283660GS KSV25F283060GN KSV25F282448GM KSV25F283672GM KSV25F283096GS KSV25F282448GS KSV25F282460GS KSV25F282448GN
Venturi V25 LX Series General Purpose Bench Fume Hoods with 28" Vertical Rising Sash, Kemglass Liner, Kewaunee Scientific
Fume Hoods and Enclosures Chemical Fume Hoods
The Supreme Air Venturi provides the safest user-friendly fume hood manufactured today.
  • Un paralleled Venturi enhanced containment technology
  • High performance/energy efficient
  • Unique ultra-smooth sash system
  • Exceptional viewing height
  • Flush airfoil
  • LED lights with color and intensity adjustability
  • Electromechanical sash stops
The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features, including: adjustable LED lights, electromechanical sash stops, ultra-smooth sash system, and push button sash management.
Certificaciones: UL and cUL listed. Tested to ASHRAE 110, NIH03-112C, ASTM D552, and NFPA 45.
Información para pedidos: Work top and base cabinets not included with fume hood.