Venturi V06 General Purpose Bench Fume Hood with Combination Vertical Rising/Horizontal Sash, Type 316L Stainless Steel Liner, Kewaunee Scientific
Supplier: Kewaunee
V06F283660LM V06F282460LM V06F282496LM V06F283696LS V06F283060LM V06F283648LM V06F282448LM V06F283096LM V06F283672LS V06F283660LS V06F283060LS V06F283672LM V06F282472LS V06F282448LS V06F283048LS V06F282460LS V06F283072LM V06F283096LS V06F283072LS V06F283696LM V06F283648LS V06F282496LS V06F283048LM V06F282472LM
KSV06F282496LMEA 28223.25 USD
KSV06F282496LM KSV06F283072LS KSV06F283060LS KSV06F282460LS KSV06F283072LM KSV06F282496LS KSV06F283060LM KSV06F283096LM KSV06F282472LS KSV06F283648LM KSV06F282448LM KSV06F282460LM KSV06F283648LS KSV06F283048LM KSV06F283096LS KSV06F282472LM KSV06F282448LS KSV06F283696LM KSV06F283696LS KSV06F283048LS KSV06F283660LS KSV06F283672LS KSV06F283672LM KSV06F283660LM
Venturi V06 General Purpose Bench Fume Hood with Combination Vertical Rising/Horizontal Sash, Type 316L Stainless Steel Liner, Kewaunee Scientific
Fume Hoods and Enclosures Chemical Fume Hoods
The Supreme Air Venturi provides the safest user-friendly fume hood manufactured today.
  • Un paralleled Venturi enhanced containment technology
  • High performance/energy efficient
  • Unique ultra-smooth sash system
  • Exceptional viewing height
  • Flush airfoil
  • LED lights with color and intensity adjustability
  • Electromechanical sash stops
The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features, including: adjustable LED lights, electromechanical sash stops, ultra-smooth sash system, and push button sash management.
Certificaciones: UL and cUL listed. Tested to ASHRAE 110, NIH03-112C, ASTM D552, and NFPA 45.
Información para pedidos: Work top and base cabinets not included with fume hood.