Anti-Eotaxin Polyclonal Antibody
10206-670
- Antibody Type:Primary
- Antigen Name:Eotaxin Picoband™
- Antigen Symbol:Eotaxin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Form:Lyophilized
- Amino Acid Number:24 to 97
- Storage Temperature:–20 °C
- Immunogen:E.coli-derived mouse Eotaxin recombinant protein (Position: H24-P97). Mouse Eotaxin shares 62% and 97% amino acid (aa) sequences identity with human and rat Eotaxin, respectively.
- Purification:purified by immunogen affinity purification
- Cat. No.:10206-670
- Supplier no.:PB9065
Polyclonal antibody for Eotaxin/CCL11 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Mouse. Eotaxin/CCL11 information: Molecular Weight: 10893 MW; Subcellular Localization: Secreted; Tissue Specificity: Expressed constitutively in the thymus. Expression inducible in the lung (type I alveolar epithelial cells), intestine, heart, spleen, kidney.
Type: Primary
Antigen: Eotaxin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse