HyperCel™ STAR AX Sorbent, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
20197-046 20197-026 20197-032
30618-492EA 2295.09 USD
30618-492 30618-488 30618-490
HyperCel™ STAR AX Sorbent, Cytiva (Formerly Pall Lab)
HyperCel™ STAR AX sorbent is designed for direct capture of proteins from bioreactor feed streams at moderate or higher salt conductivity.
  • High dynamic binding capacity (DBC) at short residence time (2 minutes or lower)
  • High flow rate properties for fast feedstock processing
Applications include direct capture of recombinant proteins, monoclonal and polyclonal antibodies, plasma derivatives or other biopharmaceuticals.