3M™ Betapure™ BK Series Filter Cartridge
Supplier: 3M Healthcare
Betapure™
BK10Z8100G1SNG BK29Z8050G2SNG BK30Z8050G1SNG BK30Z8050G1SNN BK30Z8050G1SFB BK40Z8050G1SCB BK09Z8100G1SNN BK29Z8050G2HNN BK39Z8100G2HNY
76028-553CS 856.54 USD
76028-553 76029-751 76029-773 76029-765 76029-753 76032-151 76032-179 76029-735 76032-177
3M™ Betapure™ BK Series Filter Cartridge
Filters Filter Cartridges
Betapure BK series is a truly absolute rated, rigid (non–compressible) resin bonded filter cartridge.
  • Absolute filtration efficiency at the specified reduction rating
  • Consistent production yields with absolute contaminant retention
  • No by–pass or unloading at high differential pressure
  • Choice of temperature compatible options
  • Easy disposal, suitable for incineration or shredding
  • Useable in existing filter housings with available end modifications
The absolute–rated rigid structure of Betapure™ BK series filter provides consistent performance and will not unload or lose filtration efficiency throughout its usable life!