- Home
- Water Purification Systems
- Deionizers
- 3M™ Water Softener System 3MWTS100
3M™ Water Softener System 3MWTS100
Supplier: 3M Healthcare
3M™
7100085396
76028-356EA
4215.89
USD
76028-356
3M™ Water Softener System 3MWTS100
Water Purification Systems
Deionizers
3M™ Water Softener System 3MWTS100 treats all incoming water to minimize the effects of hard water typically found in well water. Use our water softener system for brighter clothing and cleaner dishes – as well as less detergent, soap and shampoo consumption. Reduced hardness minerals throughout plumbing also decreases scale build-up in plumbing fixtures and water.
- Meets California Salt Efficiency standards for regulatory compliance
- State-of-the art water softening valve is efficient and extremely easy to use
- Superior performance brine tank features a durable, long-lasting design
- Uniquely designed salt shelf protects the valve from dirt and debris and discourages salt bridging
- Engineered system design helps provide durability and long-term operation
A User-friendly Way to Reduce Hard Water, Our 3M™ Water Softener System 3MWTS100 treats all incoming water, minimizing the effects of hard water typically found in well water applications
This system is appropriate for homes with up to three bathrooms. Water often contains calcium, magnesium and other metals that create what’s commonly known as “hard water.” These metal deposits can build up in faucets, pipes and toilets, causing equipment damage and lower water pressure. If left unaddressed, cleanup can become a major hassle and potentially quite expensive. The 3MWTS100 is an effective, user-friendly solution to this problem. 1" inlet/outlet connections, 20 – 125 psi/138 – 862 kPa pressure range, 40 – 100 °F/37.8 °C temperature range, Flow rate up to 8.9 gpm/33.7 lpm, Fully ported four position bypass valve, Meets California Salt Efficiency standards for regulatory compliance.
Brighter Clothing and Cleaner Dishes, Our 3M™ Water Softener System 3MWTS100 can remove dissolved minerals, produce brighter clothing and cleaner dishes. Treating water with a water softener system can also lead to less detergent, soap and shampoo consumption. Here’s how it works: this system saves water by metering use and regenerating only when necessary. By reducing hardness minerals throughout the plumbing system, scale build-up in plumbing fixtures and water using appliances is reduced. Additionally, the state-of-the-art valve allows monitoring and control of every function and cycle. Users enjoy the valve’s precision, which saves water and energy due to fewer regeneration cycles.
The included multi-level salt shelf keeps dirt and debris away from the brine draw area, reducing valve maintenance. The brine tank delivers superior performance by featuring a durable, long-lasting design which provides virtually trouble-free operation. Additionally, 3MWTS100 meets the California Salt Efficiency Standards (4000 grains/lb of salt). Collectively, these features are designed for long-term operation to save users time, money and energy.
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Inquire for Price
Learn more
About VWR
Item(s) Added to Cart