Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

Document Holders

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"3"
  List View Easy View
Sort by:
 
 
 
 

Catalog Number: (11005-234)

Supplier:  Production Basics
Description:   The Document Strip allows linear positioning of documents and diagrams for quick reference
Catalog Number: (11005-232)

Supplier:  Production Basics
Description:   Position documents for comfortable review with a 43.2 cm (17 in.) extension arm to allow the operator to position documents comfortably

Supplier:  Janitorial Supplies
Description:   Innovative 3-in-1 design: document holder, dry-erase board, and clipboard.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 3  of 3