Order Entry
Tungsten Point Lab Pen, Electron Microscopy Sciences
  103302-538
 :  Avantor
undefined
Tungsten Point Lab Pen, Electron Microscopy Sciences
  103302-538
 :  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Tungsten-point lab pen
  • Length:
    137 mm
  • Cat. No.:
    103302-538
  • Supplier no.:
    41148
  • Diameter:
    9.8 mm
These unique EMS pens have a sharp tungsten carbide tip which can etch glass, ceramic, and even plastic.
  • Retractable tip