The CombiFlash® Rf+ improves productivity if it is used to purify synthetic compounds, natural products, peptides or polymers. The system is compatible with both normal and reversed phase purification conditions.
- UV spectra display in real-time and post run
- RFID (Radio Frequency Identification) technology automates setting the parameters for purification runs and fraction collection
- Built in active solvent and waste management prevents columns from drying and waste overfills
- Rapid column air purge prevents solvent from being discarded with the used column
- Easy scale up with built in method scaling
- Has internal fraction collector
The TLC to Gradient calculator automatically determines optimal separation parameters while minimising run time
UV and UV-Vis detection: Variable UV 200 - 400 nm standard. Variable UV-Vis 200 - 800 nm option
Number of solvents: Binary gradient with any two of four solvents; a third solvent can be used to add modifier at a fixed percentage
Programmable gradients: Linear, step, isocratic
Sample injections: Auto-inject with 6-position automated motorised sample loading valve; self-cleans at end of run; solid or liquid injection
User interface: 10.4" touch screen
Built-in networking allows you to connect to the CombiFlash® Rf+ to monitor and remotely control your experiment, analyse data, and print to network printers.