SP Wilmad-LabGlass Pipette-Type Tilting Dispensers, SP Industries
Supplier: SP Industries
LG-7915-110 LG-7915-108 LG-7915-118
14207-678EA 425.62 USD
14207-678 14203-584 14209-214
SP Wilmad-LabGlass Pipette-Type Tilting Dispensers, SP Industries
Bottle Top Tilt Measures
These fixed volume, pipette-type dispensers are designed to deliver measured volumes quickly with reproducibility.
  • To fill, tilt backward so head is vertical
  • When assembly is returned to an upright position, excess liquid in the head flows into the flask
  • Tilting forward delivers the measured volume
Complete assembly is supplied with joint clamp (LG-1004) and flask (LG-7750).