Electrophoresis Power Supply, IBI scientific
Supplier: IBI Scientific
SH-300XL
10803-560EA 806.77 USD
10803-560
Electrophoresis Power Supply, IBI scientific
Power Supplies
Electrophoresis power supply is designed for use in the separation of DNA and RNA, blotting DNA fragmentation, immunoelectrophoresis, and protein separations.
  • Compact and lightweight
  • Ease of mobility
  • No-Load auto shut down
  • Power failure alarm with automatic re-start at current values
Electrophoresis power supply with four outputs allows the user to run four gel boxes, with the same run parameters simultaneously. Automatic cross-over function that gives the user flexibility in choosing which run parameter is best suited for the application, applied voltage, or amperage.