CONSTIX®, Sealed Polyester Swabs, SP-6N Series, Contec®
Supplier: Contec
CONSTIX®
SP-6N
76057-732CS 144.97 USD
76057-732
CONSTIX®, Sealed Polyester Swabs, SP-6N Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SP-6N sealed polyester swabs consist of 100% polyester nonwoven fabric that is thermally bonded to a flex-tip polypropylene handle.
  • Flexible tip provides a softer application on surfaces
  • Nonwoven polyester head provides enhanced sorbency for non-aqueous solutions
The polyester head is constructed without adhesives or binders, which makes the swab compatible with most common solvents, such as alcohols and ketones.