Tetrathionate Broth Base Tubed Culture Media, bioMerieux
Supplier: bioMérieux
T8034-C
95061-578CS 290.8 USD
95061-578
Tetrathionate Broth Base Tubed Culture Media, bioMerieux
Media for Microbiology Prepared Media for Microbiology
Tetrathionate Broth Base with added iodine solution is a selective enrichment medium used for the isolation of Salmonella species.
  • For the isolation of Salmonella species
  • 100 x 10 mL
Selective enrichment medium for the isolation of Salmonella species in food, also used as enrichment medium for VIDAS® SLM (with addition of iodine/brilliant green solution). Bile salts in the medium inhibit the growth of gram-positive organisms. Normal flora are inhibited by the tetrathionate formation in the medium with the addition of the iodine solution. With the selective inhibition of coliforms, the medium permits the growth of enteric pathogens virtually without any restrictions.