KISS Immersion Circulators and Circulator Baths, Huber
Supplier: Huber
2050.0004.98 2051.0004.98 2035.0013.98 2037.0041.98 2052.0004.98 2053.0004.98 2049.0004.98
76045-248EA 3944.99 USD
76045-248 76045-246 76045-250 76045-240 76045-244 76045-252 76045-242
KISS Immersion Circulators and Circulator Baths, Huber
Circulating Thermostats Closed Circulator Systems Heating Circulators
KISS circulators are ideally suited for different routine laboratory applications.
  • KISS temperature controller
  • Bright, large OLED display
  • Simple operation with menu navigation
  • Simultaneous display of setpoint, Tmin, Tmax, and actual temperature
  • Status displays for pump, cooling, and heating
  • Overtemperature and low level protection
  • USB and RS232 interfaces
  • Autostart function for power failure
KISS stands for "Keeping Innovation Safe & Simple" and describes what the customer may expect from these devices: innovative technology with safe and easy operation
The KISS range comprises an immersion circulator with screw clamp as well as different baths. The baths are available either in transparent polycarbonate (up to +100 °C) or high-grade stainless steel (up to +200 °C). The filling volume of the baths ranges from 6 to 25 L, depending on the model. For cooling applications, cooling circulators are available for working temperatures down to –30 °C. As standard, these models already work with natural refrigerants and are therefore friendly to the environment and climate.
The housing is made from high-grade stainless steel. Therefore the devices are very robust and have a very elegant appearance. However, far more important is the practicality, and KISS circulators are an excellent choice for most temperature applications. Starting with the simple commissioning, the space-saving design to the low-noise operation, KISS circulators are ideal for laboratory applications. Switch on, set setpoint value, and press start; temperature control could not be easier.
All models are equipped with an overtemperature and low level protection to class III/FL (DIN 12876) and are thus also suited for flammable liquids. Moreover, KISS circulators are a safe option from the technical application perspective.
Información accesorios: Accessories are available.