Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
Login
/
Register
Puerto Rico
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
$0.00
Order Entry
Home
D-Serine, [3-3H]
D-Serine, [3-3H] ≥97% (by HPLC), MORpure™
D-Serine, [3-3H] ≥97% (by HPLC), MORpure™
D-Serine, [3-3H] ≥97% (by HPLC), MORpure™
:
MORAVEK BIOCHEMICALS MS
D-Serine, [3-3H] ≥97% (by HPLC), MORpure™
:
MORAVEK BIOCHEMICALS MS
:
340683-22-9
:
:
2910
:
,
:
GHS07
Pack type
Size
Supplier no.
101974-006
MT944-50UCI
NextGen V-Vial
50 µCi
MT944-50UCI
Each
Each - $1,601.92
$1,601.92
-
+
Each
$1,601.92
-
+
101974-008
MT944-250UCI
NextGen V-Vial
250 µCi
MT944-250UCI
Each
Each - $2,700.38
$2,700.38
-
+
Each
$2,700.38
-
+
101974-010
MT944-1MCI
NextGen V-Vial
1 mCi
MT944-1MCI
Each
Each - $4,406.92
$4,406.92
-
+
Each
$4,406.92
-
+