BBL™ Prompt™ Inoculation System, BD
Supplier: BD Diagnostics
BBL™
226306 226306
90001-888PK 1107.77 USD
90001-888 CA-90001-888
BBL™ Prompt™ Inoculation System, BD
Spreaders Inoculating Loops and Needles
The BD BBL™ Prompt™ inoculation system is used to prepare standardized suspensions of bacteria for the Kirby-Bauer disc diffusion antimicrobial susceptibility test procedure.
The system is also used for rapidly growing bacteria such as Enterobacteriaceae, Staphylococcus spp., Pseudomonas aeruginosa, Acinetobacter spp., Enterococci , Haemophilus influenzae and some nonenterococcal streptococci.