Pierce™ Biotin Coated Plates, 96-Well Strip, Thermo Scientific
Supplier: Thermo Scientific
Pierce
15151 15151
PI15151EA 283.48 USD
CAPI15151 PI15151
Pierce™ Biotin Coated Plates, 96-Well Strip, Thermo Scientific
Microplates Immunology Microplates and Strips
Biotin coated plates can be used as the basis for a variety of biomolecular assay techniques requiring the immobilization of probes that are conjugated with streptavidin, avidin, or other biotin-binding proteins.
  • Biotin group accessible for binding avidin or streptavidin
  • Pre-blocked to reduce non specific binding
  • Strip well plate format for the ultimate in convenience
  • Coat volume: 200 µL
  • Blocking volume: 300 µL