To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:AAV Rep protein
- Clonality:Monoclonal
- Clone:259.5
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Western Blot:Yes
- Size:5 ml
- Format:Supplied in liquid form, contains 0.09% NaN3
- Storage Temperature:Store at 2-8 deg C for short term storage. Aliquot and store at -20 deg C for long term storage. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:*Blue Ice*
- Immunogen:AAV Rep protein antibody was raised in mouse using recombinant AAV-2 Rep78 protein as the immunogen.
- Cat. No.:101981-106
- Supplier no.:10R2424
Adeno-associated virus (AAV) is a small virus which infects humans and some other primate species. AAV is not currently known to cause disease and consequently the virus causes a very mild immune response. AAV can infect both dividing and non-dividing cells and may incorporate its genome into that of the host cell. These features make AAV a very attractive candidate for creating viral vectors for gene therapy, and for the creation of isogenic human disease models.
Optimal conditions to be determined by end user
Type: Primary
Antigen: AAV Rep protein
Clonality: Monoclonal
Clone: 259.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: