Anti-PUS10 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-3290
102021-548EA 718.77 USD
102021-548
Anti-PUS10 Rabbit Polyclonal Antibody
Antibodies
Pseudouridination, the isomerization of uridine to pseudouridine, is the most common posttranscriptional nucleotide modification found in RNA and is essential for biologic functions such as spliceosome biogenesis. Pseudouridylate synthases, such as PUS10, catalyze pseudouridination of structural RNAs, including transfer, ribosomal, and splicing RNAs. These enzymes also act as RNA chaperones, facilitating the correct folding and assembly of tRNAs.

WB: 1 ug/ml

Type: Primary
Antigen: PUS10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR