Anti-GTPBP2 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-1268
102017-504EA 626.46 USD
102017-504
Anti-GTPBP2 Rabbit Polyclonal Antibody
Antibodies
GTP-binding proteins, or G proteins, constitute a superfamily capable of binding GTP or GDP. G proteins are activated by binding GTP and are inactivated by hydrolyzing GTP to GDP. This general mechanism enables G proteins to perform a wide range of biologic activities.

WB: 1.25 ug/ml

Type: Primary
Antigen: GTPBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR