Anti-ARHGAP15 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-9489
102033-946EA 718.77 USD
102033-946
Anti-ARHGAP15 Rabbit Polyclonal Antibody
Antibodies
RHO GTPases regulate diverse biologic processes, and their activity is regulated by RHO GTPase-activating proteins (GAPs), such as ARHGAP15.

WB: 0.2-1 ug/ml

Type: Primary
Antigen: ARHGAP15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR