SP Wilmad-LabGlass Craig Tubes, SP Industries
Supplier: SP Industries
ML-4090-622 ML-4090-632
82003-068EA 20.13 USD
82003-068 82003-072
SP Wilmad-LabGlass Craig Tubes, SP Industries
Used for the recovery of product in microscale crystallizations, this borosilicate glass 2 ml outer tube has a non-uniform grind that purposely allows leakage (capacity is the volume below the ground area).
The PTFE plug/plunger fits outer tube. Tube and plunger sold separately.