Our MA44X series are professional clamp meters of superior quality at an affordable price.
- 1.2" (30 mm) jaw size fits conductors up to 500MCM
- Built-in non-contact voltage detector (NCV)
- Large backlit LCD display
- Functions include: NCV, AC/DC Voltage, AC current, resistance, capacitance, frequency, diode Test, continuity
- Relative mode for capacitance, data hold, auto power off and low battery indicator
Three models to choose from, with or without True RMS and AC or AC/DC Current functions. Choose True RMS model for accurate readings of non-sinusoidal waveforms, model with DC Current function for automotive applications, or model with Type K function when applications require Temperture measurements. The MA440 Series pocket-sized meters are fully loaded with multimeter functions and built-in non-contact voltage (NCV) detector to meet your application needs!
Certificaciones: CE Certified, UTL, CAT III-600, NIST Traceable.
Información para pedidos: Complete with test leads, three AAA batteries and pouch.