Vmax™ Express is a novel, fast-growing bacterial strain designed and optimized for high levels of recombinant protein expression
- Fastest growth rate of any known bacterial host with a doubling time of ~14 minutes
- Produces up to twice the biomass of E. coli with up to 4X the protein
- Enables expression of high levels of soluble recombinant proteins
- Utilizes tightly regulated, IPTG-inducible T7 promoter system
- Compatible with commonly used E. coli expression vectors, antibiotics, and growth media
- Rapid results - Go from purified plasmid to large-scale culture ready for protein purification in three days
Vmax™ cells are derived from the marine microorganism, Vibrio natriegens. This gram-negative, non-pathogenic bacterium has a doubling time twice that of E. coli and can produce greater amounts of biomass and generate large amounts of protein in a shorter time period. With a workflow similar to E. coli, this next-generation prokaryotic protein expression system can serve as a replacement for slow growing protein expression systems prone to low yields and the generation of proteins as insoluble inclusion bodies.
Designed to generate large amounts of soluble protein, Vmax Express can serve as a replacement for problematic E. coli expression systems.
Información de suministro: Shipped on dry ice. Store at ?80 °C upon arrival.