Removes excess paraffin from embedding cassettes safely and with ease by sliding them across the heated surface.
- Trims multiple blocks simultaneously
- Grooved surface
- Two temperature settings
- Includes 5 drip tray liners
The melted paraffin drains into the removable catch basin below without making a mess of paraffin shavings. Multiple blocks may be passed along the surface at the same time increasing productivity. A grooved surface allows for proper wax drainage. The trimmer features two temperature settings, low and high (approx 80 °C and approx 100 °C respectively).
The paraffin trimmer is an efficient appliance for removing excess paraffin from embedding tissue cassettes. Compact tabletop design of the paraffin trimmer fits easily in any lab. The melted paraffin is easily melted away and drains into the removable tray below. Collection tray stays in place by magnetic contact with unit. Also available is a convenient disposable tray liner that allows easy disposal of the runoff wax.
Certificaciones: CE certified and Cord is CSA and UL approved.
Información accesorios: Trimmer comes with five disposable drip tray liners. Additional liners are available as accessories.