Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biometra TOne 96 and 96G Thermal Cyclers, Analytik Jena

Supplier: Analytik Jena US

846-4-070-311 846-4-070-301 846-2-070-301 846-2-070-301 846-4-070-311 846-4-070-301 846-2-070-311 846-2-070-311
75873-970EA 5876.89 USD InStock
75873-970 CA75875-684 CA75875-688 75873-992 CA75875-864 75873-972 75873-974 CA75875-686
Biometra TOne 96 and 96G Thermal Cyclers, Analytik Jena
Thermal Cyclers
Full-featured, budget-friendly thermal cycler, integrated touchscreen control, and German-manufactured quality.

  • Optimal price/performance ratio
  • Integrated 7” touchscreen control
  • Whisper quiet: Low noise emission of max. 45 dB
  • High performance smart lid: Defined pressure control for highly reproducible results
  • Linear gradient tool: For easy gradient programming using the primer annealing temperature
  • Aluminum sample block with special alloy coating
  • Heated lid
  • Pressurized lid (manual)

Uniting Versatility and High Performance: Thanks to new electronics, the Biometra TOne can reach high heating and cooling rates of up to 4°C/sec

The Biometra TOne controls the sample block temperature without under- or overshooting the programmed target temperature. This reflects our philosophy that the instrument does exactly what the user has programmed it to do. The ingenious temperature control system incorporated into our RAC (Ramping – Accuracy – Control) technology maximizes experimental reproducibility and achieves outstanding temperature control accuracy. The Perfect Block Seal: The sample block is also perfectly sealed to prevent condensation from coming into contact with either the Peltier elements located below the sample block or with other electronic components. The seal protects the Peltier elements and extends the lifetime of the instrument.

Whisper Quiet: The airflow of the Biometra TOne thermal cycler has been optimized to keep the maximum noise level of the instrument down to an extremely low 45 decibels. This efficient airflow system also means that the Biometra TOne takes up very little space. At 26 x 43 centimeters, the dimensions of the unit would appear to be average at first glance – its effective footprint, however, also has to include the clearance zone needed to accommodate sufficient airflow at the back of the instrument. The efficient airflow of the Biometra TOne keeps this additional space requirement down to just ten centimeters – much lower than that of other thermal cyclers. A combination of low noise and a minimal footprint, Whisper Quiet technology is the result of 25 years of thermal cycler development experience.

High-Performance Smart Lid (HPSL): The heated lid of the Biometra TOne is equipped with HPSL technology. An integrated slip clutch always maintains constant contact pressure, regardless of the shape and height of the plasticware. This optimizes the contact between the sample block well and the walls of the plasticware, resulting in reproducible conditions. As soon as the heated lid is closed, a rubber seal on the lid encapsulates the space surrounding the sample block. This closed space increases the sample block temperature uniformity and prevents condensation formation during the final PCR cooling step.

The intelligent way to program a gradient: Determining the optimum annealing temperature is a challenge when creating a new primer pair. Using the gradient tool allows the operator to find the optimum annealing temperature under experimental conditions and apply that temperature value to routine applications. Programming gradients usually means setting temperature for the first and the last row to determine the gradient range. For most thermal cyclers, the temperature interval is not the same from one row of the sample block to the next. In addition, the sigmoidal temperature curve of the gradient brings two disadvantages: 1. The automatically resulting temperatures for the other rows of the sample blocks, in particular in its center, are almost always odd-numbered. 2. The temperature differences between the rows are unevenly distributed, increasing towards the center of the sample block.
The Linear Gradient Tool (LGT) enables programming of a temperature gradient with defined temperature intervals (increment) between the individual rows of the sample block. It allows the user to enter the calculated Tm value (minus e.g. 5°C) and the desired temperature increment (e.g. ± 1 °C) between the rows of the sample block. For maximum comfort, the Linear Gradient Tool now supports programming of even-numbered temperature values for a maximum number of rows.
Pre-installed program templates: The Biometra TOne software offers a number of preinstalled program templates for different applications. The program templates provide a general protocol structure and can be easily adapted for the current experiment.
Multi-step programming: Editing a template or setup of new protocols is quite simple due to the multi-step programming feature, which allows users to enter all of the parameters for every program step within a single screen. Thus, the multi-step programming renders constantly switching back and forth between different screens unnecessary and enables easy changes from step to step.

Información para pedidos: Two-year warranty.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty