Silver Talon® MasterGrip® Gloves with High Grade Leather Palm and Fingers, Wells Lamont
Supplier: Wells Lamont
Silver Talon®
134778 134775 134772 134779 134770 134767 134769 134776 134766 134773
80090-711EA 68 USD
80090-711 80090-699 80090-713 80090-701 80090-715 80090-703 80090-705 80090-707 80090-709 80090-697
Silver Talon® MasterGrip® Gloves with High Grade Leather Palm and Fingers, Wells Lamont
Gloves
Silver Talon® MasterGrip® gloves with high grade leather on palms and fingers for abrasion, cut, heat and some puncture resistance.
  • Based on Silver Talon® construction with added leather on palm and fingers
  • Abrasion, cut, heat and some puncture resistance
  • Features Honeywell Spectra® Fiber
  • High grade leather palm patch
  • Leather palm absorbs oil and grease, increasing grip capabilities
Silver Talon® MasterGrip® gloves are available in hand-specific sizes. They feature Honeywell Spectra® Fiber. These gloves are offer abrasion, cut, heat and some puncture resistance. High-grade leather on the palms and fingers of these gloves absorbs oil and grease, and it increases the wearer's ability to grip materials.
Certificaciones: ANSI Level 5, A6.
Precaución: WARNING: Cut resistant gloves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades.