Slipguard® Heavy Duty Cut Resistant Standard Pattern Gloves, Wells Lamont
Supplier: Wells Lamont
Slipguard®
133804 133787 133799 134157 133792 133796 133795 133788 133803 134156 133800 133791
87000-563EA 61.4 USD
87000-563 87000-421 14231-977 87000-419 87000-417 14231-973 87000-415 14231-975 87000-425 87000-565 14231-971 87000-423
Slipguard® Heavy Duty Cut Resistant Standard Pattern Gloves, Wells Lamont
Gloves
Slipguard® prewashed cut-resistant gloves with industry standard, slip-resistant pattern.
  • Provides cut and slip protection
  • Industry leading Handguard II construction with polyurethane grip pattern
  • Features Honeywell Spectra® Fiber
  • Standard pattern is ideal for use in plastic trimming
  • Prewashed
  • Preshrunk
Slipguard® heavy duty gloves come in hand-specific sizes. They are made out of Honeywell Spectra® Fiber and feature an industry-leading Handguard II construction with a polyurethane grip pattern. This standard pattern makes these gloves ideal for work with plastics. These gloves offer the ideal balance between cut protection and slip resistance. Gloves are prewashed and preshrunk for a comfortable fit.
Certificaciones: ANSI Cut Level 5, A8.
Precaución: WARNING: Cut resistant gloves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades.