VeriSpec® Mixed Anion Standard, 7 Component, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010687-100N
RCRV010687100NEA 396.51 USD
RCRV010687100N
VeriSpec® Mixed Anion Standard, 7 Component, Ricca Chemical Company
Standards Anionic Standards Multi-Element Anioinic Standards
Certified reference material (CRM) for ion chromatography (IC)
200 ppm PO₄³; 100 ppm SO₄²-, NO₃-, Br-, NO₂-, Cl-; 20 ppm F-
Certificaciones: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility