6-Well Plate with Lid, Polystyrene, Greiner Bio-One
Supplier: Greiner bio-one
657102
490007-412CS 142.01 USD
490007-412
6-Well Plate with Lid, Polystyrene, Greiner Bio-One
Microplates Cell Culture Plates
6-Well Macroplates are manufactured from crystal-clear and heat-resistant polystyrene for optimal analysis by microscopy.
  • With 35 mm diameter per well, it is a practical and space-saving alternative to six standard petri dishes
  • Numeric coding allows easier identification of the wells
  • With vents
  • Single position lid with condensation rings
  • Nonsterile
Durable polystyrene construction and clear siding allow for safe handling of easily observable samples. Macroplates are available in a variety of formats to allow multiple growth experiments to be run simultaneously.