Pressure Reaction Bottles, Wilmad-Labglass
Supplier: Wilmad-LabGlass
LG-3921-104 LG-3921-116 LG-3921-110 LG-3921-100 LG-3921-108 LG-3921-114 LG-3921-106 LG-3921-102 LG-3921-112
14211-532EA 64.26 USD
14211-532 14208-144 14212-742 14201-480 14202-280 14204-280 14210-916 14211-404 14201-790
Pressure Reaction Bottles, Wilmad-Labglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Bottles are widely used in pressure reaction studies.
  • Fabricated from selected heavy wall borosilicate glass
  • Pressure rating of 55 lbs