Alltech® 3300 ELSD, BUCHI
Altech
145135834
75841-102EA 26549.8 USD
75841-102
Alltech® 3300 ELSD, BUCHI
The Alltech® ELSD simplifies analysis of difficult compounds such as pharmaceuticals, impurities, fatty acids, polymers and more.
  • ELSD improves baseline stability and detection sensitivity compared to refractive index (RI)
  • ELSD shows what may be missing from your UV chromatogram
  • Use ELSD in parallel with UV/Vis and MS for maximum structural and concentration information
  • ELSD obtains a more accurate representation of sample mass than UV/Vis
Certificaciones: CSA listed, complies with 21 CFR Subpart J