400 A AC/DC TRMS Clamp Meter, Type K Thermocouple, Extech
Supplier: Extech
CM46
75839-410EA 444.45 USD
75839-410
400 A AC/DC TRMS Clamp Meter, Type K Thermocouple, Extech
Electrical Meters Digital Multimeters
Professional 400A AC/DC True RMS Clamp Meter with Accu-Tip and Type K Thermocouple
Black color.
Certificaciones: UL , CE.
Información para pedidos: Complete wth? TA80 test lead set, carrying pouch with FLIR logo, CD manual, 2x AAA Engergizer batteries. Type K thermocouple.