innuPREP PCRpure 96 Kit, Analytik Jena
Supplier: Analytik Jena
845-FP-5010384 845-FP-5010192 845-FP-5010960 845-FP-5010192 845-FP-5010384
10755-774PK 452.02 USD
10755-774 10755-776 CA10755-774 CA10755-776 CA10755-778
innuPREP PCRpure 96 Kit, Analytik Jena
Protein Purification Protein Purification Kits
The innuPREP PCRpure Kit provides an extremely fast, simple, and highly efficient method for purifying amplification products directly from PCR reaction mixtures and/or for concentrating PCR products.
  • A new, 2-step process for performing PCR purification in just 3 minutes
  • Ability to process very small elution volumes of at least 10µl
  • High rates of recovery for a large range of fragment lengths
  • Extremely fast and simple with minimal steps involved
The innuPREP PCRpure Kit provides an extremely fast, simple, and highly efficient method for purifying amplification products directly from PCR reaction mixtures and/or for concentrating PCR products. Purification is based on a two-step method and takes only approx. 3 minutes to complete. The need for previously-standard wash steps is eliminated, thereby reducing the overall process to binding and elution. The process makes it possible to recover amplification products ranging in size from 60 bp to 30 kb with recovery rates of 75% to 95%, depending on the length of the amplification product. Also, elution can be performed with a very small volume of just 10µl, which eliminates the need for specialized “mini-elute” spin filter columns.