Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
Login
/
Register
Puerto Rico
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
$0.00
Order Entry
AICA ribonucleotide
Supplier:
Avantor
Certificates
Expand image
AICA ribonucleotide
Supplier:
Avantor
About this item
:
3031-94-5
:
:
C₉H₁₅N₄O₈P
:
MFCD00057264
:
338.213 g/mol
:
3316
:
,
Related Information
Documentation
9416.pdf
ghs_9416.pdf