ExoStd™ Lyophilized Exosome Standard, 30 µg, Human Plasma, BioVision Inc
Supplier: BioVision
M1040-6
75799-568EA 1342.63 USD
75799-568
ExoStd™ Lyophilized Exosome Standard, 30 µg, Human Plasma, BioVision Inc
Lyophilized and purified Exosomes available from Human Plasma (30 µg).
  • Purified and lyophilized exosomes
  • Control standards for multiple applications including FACS, WB, ELISA
  • Calibration standards for quantitation of exosome-derived markers from biological samples
  • Easy to ship and store
  • Stable for over 36 months at 4°C