The MCP 300 is an automatic polarimeter for analyzing optically active substances
- High accuracy over the entire measuring range
- Toolmaster™ - Wireless automatic identification of sample cells and quartz control plates via RFID
- Modular concept
- Automatic Peltier temperature control
- LED light source – no maintenance for 100,000 hours
- Up to eight spectral filters in the UV/VIS/NIR ranges (optional)
- Full international compliance
- International and national pharmacopeias
- 21 CFR Part 11
- OIML
- Full QM compliance
- Password protection with three user levels
- Audit trail
- GMP/GLP compatible
- Forgery-proof data export
The combination of quality, robust design and modern data management makes MCP 300 the most powerful polarimeter on the market. The instrument allows customer-specific configurations and provides the widest range of options for the future. The MCP 300 meets all application requirements for the pharmaceutical, cosmetics, chemical and medical industries as well as for R&D. Multi-Wavelength module for up to eight wavelengths features a filter wheel that allows for the installation of up to eight different interference filters to measure at different wavelengths.
It is highly accurate and easy to use.
Certificaciones: CE Compliant.
Información para pedidos: An instruction manual, power cord, two replacement fuses, CAN open male termination plug and CAN open female termination plug are supplied with every instrument.
Every MCP 300 needs to be ordered together with at least one sample cell and a quartz control plate. Many additional sample cells and quartz control plates not listed are available by request.