United States
Order Entry
Arsenic ICP Standard, Inorganic Ventures
 
undefined
Arsenic ICP Standard, Inorganic Ventures
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Environmentally Preferable:

 

 

  •  
     
    Element
    Concentration
    Size
    Color
    Matrix
    Supplier no.
     
     
  • 101372-438
    CGAS(5)1-125ML
    Arsenic
    1000 µg/ml
    125 ml
    Clear
    H₂O
    CGAS51-1
       
       
    Each (125ml)
    Each (125ml) - $59.23
    $59.23
     
    Each (125ml)
       $59.23

  • 100360-574
    CGAS1-500ML
    Arsenic
    1000 µg/ml
    500 ml
    Clear
    2% HNO₃
    CGAS15
       
       
    Each (500ml)
    Each (500ml) - $123.85
    $123.85
     
    Each (500ml)
       $123.85

  • 101372-440
    CGAS(5)1-500ML
    Arsenic
    1000 µg/ml
    500 ml
    Clear
    H₂O
    CGAS51-5
       
       
    Each (500ml)
    Each (500ml) - $123.85
    $123.85
     
    Each (500ml)
       $123.85

  • 101372-434
    CGAS(3)1-125ML
    Arsenic
    1000 µg/ml
    125 ml
    Clear
    HCl/NaOH/NaHCO₃
    CGAS31-1
       
       
    Each (125ml)
    Each (125ml) - $59.23
    $59.23
     
    Each (125ml)
       $59.23

  • 101372-436
    CGAS(3)1-500ML
    Arsenic
    1000 µg/ml
    500 ml
    Clear
    HCl/NaOH/NaHCO₃
    CGAS31-5
       
       
    Each (500ml)
    Each (500ml) - $123.85
    $123.85
     
    Each (500ml)
       $123.85

  • 100359-874
    CGAS10-500ML
    Arsenic
    10000 µg/ml
    500 ml
    Clear
    2% HNO₃
    CGAS105
       
       
    Each (500ml)
    Each (500ml) - $217.54
    $217.54
     
    Each (500ml)
       $217.54