You Searched For: Ruthenium Standards


13  results were found

SearchResultCount:"13"

Sort Results

List View Easy View

Rate These Search Results

Supplier: BDH
Description: Used with ICP-MS, ICP-OES, DCP, GFAA, and AA
Supplier: Thermo Scientific Chemicals
Description: AAS standard Solution, Specpure reagent, Ru 1000microg/mL.100mL.
Supplier: Thermo Scientific Chemicals
Description: plasma standard solution, Specpure*, Ru 10,000æg/ml,UN1789,Liquid,WARNING: CORROSIVE, irritates skin and eyes,100ml,500ml,CORROSIVE,NO CAN FOR GROUND SHIPMENT
Catalog Number: (RCARU1KH-100)
Supplier: Ricca Chemical
Description: 1000 ppm Ru ion in dilute HCl. For use in standardizing atomic absorption spectrophotometers. 100mL.

Supplier: Thermo Scientific Chemicals
Description: plasma standard solution, Specpure*, Ru 1000æg/ml,UN1789,Liquid,WARNING: CORROSIVE, irritates skin and eyes,100ml,500ml,CORROSIVE,NO CAN FOR GROUND SHIPMENT
Supplier: Ricca Chemical
Description: VeriSpec* Ruthenium (Ru) Standard for AAS 1000ppm in 2% HCl, Form: Liquid, Container: Poly natural, Manufactured & Tested in ISO 17025/Guide 34 Accredited Facility, NIST traceable with specification tolerances ensu

Supplier: Ricca Chemical
Description: VeriSpec* Ruthenium Standard, For ICP/MS 100 ppm in 2% HCl Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility, Form: Liquid, Container: Poly natural, NIST traceable, Size: 100ML

Supplier: Ricca Chemical
Description: 1000ppm Ru. Single element inductively coupled plasma (ICP) metal standard for use in standardizing ICP and AA spectrophotometers for parts per million (ppm) and parts per billion (ppb) analyses. 500mL.
Supplier: Ricca Chemical
Description: VeriSpec* Ruthenium Standard, For ICP 1000 ppm in 5% HCl Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility, Form: Liquid, Container: Poly natural, NIST traceable, Size: 500ML

Supplier: SPEX CERTIPREP LLC
Description: Single-element standards for AA & ICP.

Catalog Number: (BT122995-50ML)
Supplier: BeanTown Chemical
Description: EC No: 231-595-7; MDL No: MFCD00011207

SDS


Catalog Number: (BT134875-500ML)
Supplier: BeanTown Chemical
Description: EC No: 231-595-7; MDL No: MFCD00011207

SDS


Catalog Number: (100360-642)
Supplier: Inorganic Ventures
Description: Certified reference material for Ruthenium, traceable to NIST.


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 13 of 13
no targeter for Bottom