Fluorometric NADH Quantitation Kit, PromoKine, PromoCell
Supplier: PromoCell
PK-CA577-K338
10183-722EA 812.8 USD
10183-722
Fluorometric NADH Quantitation Kit, PromoKine, PromoCell
Assays Enzyme Assays
PromoKine’s Fluorometric NADH Quantitation Kit provides a highly sensitive method to detect very low level of NADH.
  • Can be used for studies of energy transformation
  • Determination of the redox state of cells or tissue
  • Designed to specifically recognize NADH
PromoKine’s Fluorometric NADH Quantitation Kit provides a highly sensitive method to detect very low level of NADH (while NAD are not detected in the reaction). It can be used for studies of energy transformation and the determination of the redox state of cells or tissue. The kit has been designed to specifically recognize NADH only in an enzyme cycling reaction that significantly increases detection sensitivity.