QuantiTect Probe RT-PCR kit for one-step qRT-PCR using sequence-specific probes for gene expression analysis.
- Sensitive detection of as few as 10 copies of each target
- Accurate quantification
- Use of any sequence-specific probe on any real-time cycler
- No need to optimize reaction and cycling conditions
Includes 3 x 1.7mL 2x QuantiTect Probe RT-PCR Master Mix, 100L QuantiTect RT Mix, and 2 x 2mL RNase-free water.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Application:RT-qPCR probe
- No. of reactionsSupplier no.
- QuantiTect Probe RT-PCR Kit
- QuantiTect Probe RT-PCR Kit