DuPont™ Tychem® 6000 FR Bib Coverall and Jacket Combo
Supplier: DuPont
Tychem®
TP750TORMD000200 TP750TORXL000200 TP750TOR4X000200 TP750TOR3X000200 TP750TORLG000200 TP750TOR2X000200 TP750TORSM000200
10016-880CS 1401.89 USD
10016-880 10016-882 10016-884 10016-886 10016-876 10016-888 10016-878
DuPont™ Tychem® 6000 FR Bib Coverall and Jacket Combo
Disposable Garments
Tychem® 6000 FR offers a single layer garment that provides triple hazard protection from liquid-chemical splash, flash fire and electric arc.
  • Bib overall and jacket combo
  • Elastic at wrists and bottom of jacket
  • Open ankles
  • Double storm flap on jacket
  • Adjustable webbing straps with buckle closure on overalls
  • Taped seams
Tychem® 6000 FR garments are designed to combine the trusted chemical protection of Tychem® and thermal/arc protection of Nomex® into a single layer garment. Tychem® 6000 FR garments are designed to help provide industrial workers and Hazmat responders with permeation protection against a broad range of toxic industrial chemicals and chemical warfare agents and to provide escape time from flash fires and protection from electric arc. Tychem® 6000 FR fabric provides at least eight hours breakthrough protection against 17 of the 21 standard ASTM F1001 chemicals and at least 30 minutes of protection for more than 180 chemical challenges including chemical warfare agents. The arc rating of Tychem® 6000 FR fabric is 15 cal/cm² Ebt. This exceeds the NFPA 70E Hazard Risk Category 2 requirement of 8 cal/cm². Typical Applications include HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories.
Garments are sewn with Nomex® thread and over-taped with chemical barrier tape of same flame resistant composition as garment fabric. Key garment components (fabric, zipper, drawstrings) are flame resistant and thermally stable (will remain functional even after 5 minutes exposure to 260°C). Garments were developed to meet the needs of industrial plant sites enabling easy donning and doffing.
Certificaciones: Thickness meets ASTM D1117. Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Información para pedidos: Jacket and bib overall combination ensemble are sold together. Jacket and bib overall not sold separately.