DuPont™ Tychem® 6000 FR Coveralls with Respirator Hood, Certified to NFPA 1992 and NFPA 2112
Supplier: DuPont®
Tychem®
TP198TGYLG000200 TP198TGY2X0002BN TP198TORLG0002BN TP198TOR5X0002BN TP198TOR5X000200 TP198TGYLG0002BN TP198TGY5X0002BN TP198TGY2X000200 TP198TORLG000200 TP198TOR2X0002BN TP198TOR2X000200 TP198TGYSM0002BN TP198TGY4X000200 TP198TGY5X000200 TP198TOR3X000200 TP198TOR4X0002BN TP198TGYMD000200 TP198TORSM0002BN TP198TORMD0002BN TP198TGYXL000200 TP198TORMD000200 TP198TGYSM000200 TP198TGY4X0002BN TP198TGYMD0002BN TP198TGY3X000200 TP198TOR4X000200 TP198TORSM000200 TP198TORXL0002BN TP198TGYXL0002BN TP198TOR3X0002BN TP198TGY3X0002BN TP198TORXL000200
10016-590CS 1282.39 USD
10016-590 10016-576 10016-624 10016-620 10016-618 10016-592 10016-588 10016-574 10016-622 10016-608 10016-606 10016-600 10016-582 10016-586 10016-610 10016-616 10016-594 10016-632 10016-628 10016-602 10016-626 10016-598 10016-584 10016-596 10016-578 10016-614 10016-630 10016-636 10016-604 10016-612 10016-580 10016-634
DuPont™ Tychem® 6000 FR Coveralls with Respirator Hood, Certified to NFPA 1992 and NFPA 2112
Disposable Garments
Tychem® 6000 FR offers a single layer garment that provides triple hazard protection from liquid-chemical splash, flash fire and electric arc.
  • Certified to NFPA 1992, NFPA 2112, and the Category 2 Requirements of NFPA 70E
  • Respirator fit hood with drawstring
  • Elastic wrists
  • Open ankles
  • Double storm flap with hook & loop closure
  • Taped seams
Tychem® 6000 FR garments are designed to combine the trusted chemical protection of Tychem® and thermal/arc protection of Nomex® into a single layer garment. Tychem® 6000 FR garments are designed to help provide industrial workers and Hazmat responders with permeation protection against a broad range of toxic industrial chemicals and chemical warfare agents and to provide escape time from flash fires and protection from electric arc. Tychem® 6000 FR fabric provides at least eight hours breakthrough protection against 17 of the 21 standard ASTM F1001 chemicals and at least 30 minutes of protection for more than 180 chemical challenges including chemical warfare agents. The arc rating of Tychem® 6000 FR fabric is 15 cal/cm² Ebt. This exceeds the NFPA 70E Hazard Risk Category 2 requirement of 8 cal/cm². Typical Applications include HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories.
Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Attached respirator-fit hood features a drawstring to pull tightly around respirator. A longer zipper extends to chin for complete coverage of neck area. Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Elastic openings at wrists allow for a tighter fit. Open ankles are hemmed.
Certificaciones: Thickness meets ASTM D1117. Certified to NFPA 1992 (2012) and NFPA 2112 (2012). Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant.