Acetonitrile, anhydrous ≥99.9% for DNA synthesis
Supplier: VWR International
Synonyms:
Cyanomethane
From the exclusive family of high-quality, competitively priced BDH Chemicals – High Purity solvents for HPLC applications.
Solvents are pumped through 0.2 micron filters on filling and packaged under Nitrogen.
Envase: All bottles have tamper evident caps for added peace of mind and are packed in environmentally –friendly corrugated/pulp case inserts that are fully recyclable.
Formula:
H₃CCN MW: 41.05 g/mol Boiling Pt: 81.6 °C (1013 hPa) Melting Pt: –45.7 °C Density: 0.782 g/cm³ (20 °C) Flash Pt: 2 °C (closed cup) |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 UN: 1648 ADR: 3,II |
Specification Test Results
Assay | Min. 99.9% |
IR Spectrum | Passes test |
Acidity | Max. 0.001 meq/g |
Alkalinity | Max. 0.0002 meq/g |
Coloration | Max. 10 APHA |
Evaporation residue | Max. 1 ppm |
Water | Max. 10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...