A range of eco-friendly, refrigerated/heating circulating baths with an intuitive user interface and sleek design.
- Full colour 5 language QVGA TFT display on both the ecocool 150 and ecocool 200 units
- Active cooling through the whole temperature range up to 200°C
- eco mode optimises the cooling control needed, enabling rapid cool down when required
- True energy saving of up to 80% in idle state compared with standard compressor chillers which only switch the compressor off
- Thermostat and chiller work in harmony, neither will operate alone, eliminating any danger of overheating or freezing
- No-spill valved front drain and single front switch for user convenience
- No side vents, locate to suit the user, not the unit; Thermostat can be positioned at 90° for length way position
Units offer significant running cost savings whilst delivering powerful cooling when needed. Choice of three models with temperature range of -25 to 200°C (model dependent*). Ideal for applications in pharmaceutical, education, industrial and life science areas for use with mini pilot plant reactors, cooling rotary evaporators, replacing running tap water cooling, immersing small samples, photometry, chromatography systems, sample preparation, general cooling, reaction chemistry, temperature control, semi-conductor manufacturing, rheometry, refractometry and electrophoresis cooling.
LT ecocool™100 is a general purpose circulating bath with digital display for use from -20 to 100°C, delivers 250W of cooling power.
LT ecocool™150 is a general purpose circulating bath with digital display for use from -25 to 150°C, delivers 350W of cooling power. Unit is programmable (1 x 30) with adjustable pump, USB and temperature probe socket.
Información para pedidos: All models are supplied assembled as ready to use kits, complete with accessory hosing*, clips and connectors as standard. Optional IQ/OQ/PQ documentation, high temperature hosing, high pressure pumps, alternative connectors, temperature probes and Labwise™ control software (for LT ecocool™ 150 and 200) are also available.