Puerto Rico
Order Entry
Anti-IgD Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: IA6-2]
Anti-IgD Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: IA6-2]
 :  Biolegend
 
Anti-IgD Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: IA6-2]
 :  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 348217-BL
  • 348218-BL
  • Antigen Symbol:
  • IgD
  • IgD
  • Antigen Synonyms:
  • Ig delta chain C region
  • Ig delta chain C region
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)/Cy7®
  • APC (Allophycocyanin)/Cy7®
  • Clone:
  • IA6-2
  • IA6-2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3495
  • 3495
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Immunogen:
  • Human IgD
  • Human IgD
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with APC/Cy7 under optimal conditions. The solution is free of unconjugated APC/Cy7 and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with APC/Cy7 under optimal conditions. The solution is free of unconjugated APC/Cy7 and unconjugated antibody.
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

 

 

  •  
     
    Size
    Supplier no.
     
     
  • 348217-BL
    348217
    25 tests
    348217
       
       
    Each
    Each - $206.42
    $206.42
    Each
       $206.42

     
     
     
    Specifications
    Antigen Symbol
    IgD
    Environmentally Preferable
    Antigen Synonyms
    Ig delta chain C region
    Antibody Type
    Primary
    Clonality
    Monoclonal
    Conjugation
    APC (Allophycocyanin)/Cy7®
    Clone
    IA6-2
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    3495
    Isotype
    IgG2a kappa
    Immunogen
    Human IgD
    Form
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
    Purification
    The antibody was purified by affinity chromatography and conjugated with APC/Cy7 under optimal conditions. The solution is free of unconjugated APC/Cy7 and unconjugated antibody.
    Storage Temperature
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
    Shipping Temperature
    Ambient RT
  • 348218-BL
    348218
    100 tests
    348218
       
       
    Each
    Each - $521.22
    $521.22
    Each
       $521.22

     
     
     
    Specifications
    Antigen Symbol
    IgD
    Environmentally Preferable
    Antigen Synonyms
    Ig delta chain C region
    Antibody Type
    Primary
    Clonality
    Monoclonal
    Conjugation
    APC (Allophycocyanin)/Cy7®
    Clone
    IA6-2
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    3495
    Isotype
    IgG2a kappa
    Immunogen
    Human IgD
    Form
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
    Purification
    The antibody was purified by affinity chromatography and conjugated with APC/Cy7 under optimal conditions. The solution is free of unconjugated APC/Cy7 and unconjugated antibody.
    Storage Temperature
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
    Shipping Temperature
    Ambient RT