Puerto Rico
Order Entry
Anti-CD73 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: TY/11.8]
Anti-CD73 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: TY/11.8]
 :  Biolegend
 
Anti-CD73 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: TY/11.8]
 :  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 127211-BL
  • 127212-BL
  • Antigen Symbol:
  • CD73
  • CD73
  • Antigen Synonyms:
  • NT5E|Ecto-5’-nucleotidase
  • NT5E|Ecto-5’-nucleotidase
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PB (Pacific Blue)
  • PB (Pacific Blue)
  • Clone:
  • TY/11.8
  • TY/11.8
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 23959
  • 23959
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Immunogen:
  • CHO cells transfected with mouse CD73
  • CHO cells transfected with mouse CD73
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™.
  • The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™.
  • Concentration:
  • 0.50 mg/mL
  • 0.50 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

 

 

  •  
     
    Size
    Supplier no.
     
     
  • 127211-BL
    127211
    25 μg
    127211
       
       
    Each (25µG)
    Each (25µG) - $139.60
    $139.60
    Each (25µG)
       $139.60

     
     
     
    Specifications
    Antigen Symbol
    CD73
    Environmentally Preferable
    Antigen Synonyms
    NT5E|Ecto-5’-nucleotidase
    Antibody Type
    Primary
    Clonality
    Monoclonal
    Conjugation
    PB (Pacific Blue)
    Clone
    TY/11.8
    Reactivity
    Mouse
    Host
    Rat
    Gene ID
    23959
    Isotype
    IgG1 kappa
    Immunogen
    CHO cells transfected with mouse CD73
    Form
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
    Purification
    The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™.
    Concentration
    0.50 mg/mL
    Storage Temperature
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
    Shipping Temperature
    Ambient RT
  • 127212-BL
    127212
    100 μg
    127212
       
       
    Each (100µG)
    Each (100µG) - $298.30
    $298.30
    Each (100µG)
       $298.30

     
     
     
    Specifications
    Antigen Symbol
    CD73
    Environmentally Preferable
    Antigen Synonyms
    NT5E|Ecto-5’-nucleotidase
    Antibody Type
    Primary
    Clonality
    Monoclonal
    Conjugation
    PB (Pacific Blue)
    Clone
    TY/11.8
    Reactivity
    Mouse
    Host
    Rat
    Gene ID
    23959
    Isotype
    IgG1 kappa
    Immunogen
    CHO cells transfected with mouse CD73
    Form
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
    Purification
    The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™.
    Concentration
    0.50 mg/mL
    Storage Temperature
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
    Shipping Temperature
    Ambient RT